Learn More
Keystone Cleanroom Products Blue SMS Sterilization Wrap, 50 gsm
SMS sterilization wraps provide a microbial barrier and are ideal for pharmaceutical production applications. They are designed for use in steam, ethylene oxide, and hydrogen peroxide sterilization processes.
266.00 € - 1260.00 €
Specifications
Product Type | Sterilization Wrap (Heavy-duty) |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | COC |
Closure Type | None |
Color | Blue |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
17851471
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-01.5X01.5 |
3.8 x 3.8 cm (1.5 x 1.5 in.) |
405.00 €
Pack of 3000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17861471
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0202 |
5.1 x 5.1 cm (2 x 2 in.) |
405.00 €
Pack of 3000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17871471
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0303 |
7.6 x 7.6 cm (3 x 3 in.) |
405.00 €
Pack of 3000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17881471
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0404 |
10.2 x 10.2 cm (4 x 4 in.) |
405.00 €
Pack of 3000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17891471
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0606 |
15.2 x 15.2 cm (6 x 6 in.) |
405.00 €
Pack of 3000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17801481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0808 |
20.3 x 20.3 cm (8 x 8 in.) |
278.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17811481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0909 |
22.9 x 22.9 cm (9 x 9 in.) |
290.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17821481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-0912 |
22.9 x 30.5 cm (9 x 12 in.) |
371.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17831481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-1212 |
30.5 x 30.5 cm (12 x 12 in.) |
402.48 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17841481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-1515 |
38.1 x 38.1 cm (15 x 15 in.) |
464.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17851481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-1818 |
45.7 x 45.7 cm (18 x 18 in.) |
651.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17861481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-1822 |
45.7 x 55.9 cm (18 x 22 in.) |
1260.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17871481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-2020 |
50.8 x 50.8 cm (20 x 20 in.) |
754.00 €
Pack of 1000 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17881481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-2424 |
61 x 61 cm (24 x 24 in.) |
266.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17891481
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-3030 |
76.2 x 76.2 cm (30 x 30 in.) |
454.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17801491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-3636 |
91.4 x 91.4 cm (36 x 36 in.) |
636.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17811491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-4040 |
101.6 x 101.6 cm (40 x 40 in.) |
805.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17821491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-4545 |
114.3 x 114.3 cm (45 x 45 in.) |
964.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17831491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-4548 |
114.3 x 121.9 cm (45 x 48 in.) |
1090.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17841491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-4848 |
121.9 x 121.9 cm (48 x 48 in.) |
1090.00 €
Pack of 250 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17851491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-5454 |
137.2 x 137.2 cm (54 x 54 in.) |
464.00 €
Pack of 100 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17861491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-5472 |
137.2 x 182.9 cm (54 x 72 in.) |
723.00 €
Pack of 100 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17871491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-6060 |
152.4 x 152.4 cm (60 x 60 in.) |
764.00 €
Pack of 100 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
17881491
![]() ![]() |
Keystone Cleanroom Products
KCRP-300-6080 |
152.4 x 203.2 cm (60 x 80 in.) |
405.00 €
Pack of 50 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
Description
- Manufactured and packaged in a Certified ISO 8 cleanroom
- Clean - no cellulose
- Strong, rip and tear resistant 50 gsm material
- Soft and drape-able fabric
- An extensive range of sizes available means less cutting
- Hydrophobic barrier allows sterilization gasses but prevents liquid and microbial penetration
- Sub packed into sealed labeled poly bags with Lot Number and Date of Manufacture for complete traceability
- Certificate of Conformance sent with each shipment
- Made in the USA
Specifications
Sterilization Wrap (Heavy-duty) | |
COC | |
Blue | |
SMS Fabric | |
Yes |
Autoclavable | |
None | |
Single-use | |
Non-sterile | |
121°C to 125°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Keystone Cleanroom Products Blue SMS Sterilization Wrap, 50 gsm