Learn More
Bohlender™ BOLA™ Spherical Ground Joint Sleeves
Spherical ground joint sleeves with gripping ring and sealing rings on the outside.
Brand: Bohlender™ H934-12
This item is not returnable.
View return policy
Description
- With gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.01 mbar x l x s-1
- Solid construction for continued use
Specifications
PTFE | |
Ground Joint Sleeves | |
S 40 |