Learn More
Bohlender™ BOLA™ Spherical Ground Joint Sleeves
Spherical ground joint sleeves with gripping ring and sealing rings on the outside.
21.00 € - 36.00 €
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
17137404
|
Bohlender™
H934-02 |
S 13 |
23.00 €
Pack of 3 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
17147404
|
Bohlender™
H934-04 |
S 19 |
21.00 €
Pack of 3 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
17157404
|
Bohlender™
H934-06 |
S 29 |
25.00 €
Pack of 3 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
17177404
|
Bohlender™
H934-12 |
S 40 |
36.00 €
Pack of 2 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
Description
- With gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.01 mbar x l x s-1
- Solid construction for continued use
Specifications
PTFE | |
Ground Joint Sleeves |