missing translation for 'onlineSavingsMsg'
Learn More

Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 175081000


 View more versions of this product

Product Code. 10781641

  • 51.50 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borane-tetrahydrofuran complex
1M solution in tetrahydrofuran, stabilized
C4H11BO
0.96 to 1.08M
0.8760g/mL
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
RMCYTHFAWCWRFA-UHFFFAOYSA-N
oxolane borane
11062302
AcroSeal™ Glass bottle
Stabilized
−22°C
Colorless
14044-65-6, 109-99-9
MFCD00012429
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101
15, 1336
Solubility in water: reacts.
B.C1CCOC1
85.94
85.94
0.876
100 mL
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Borane-tetrahydrofuran complex, 1M solution in THF, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, Borane-tetrahydrofurancomplex

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only