Learn More
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, Thermo Scientific Chemicals
51.50 € - 384.00 €
Specifications
Chemical Name or Material | Borane-tetrahydrofuran complex |
---|---|
Color | Colorless |
Name Note | 1M solution in tetrahydrofuran, stabilized |
CAS | 14044-65-6, 109-99-9 |
Molecular Formula | C4H11BO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10781641
|
Thermo Scientific Acros
175081000 |
100 mL | AcroSeal™ Glass bottle |
51.50 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781831
|
Thermo Scientific Acros
175088000 |
800 mL | AcroSeal™ Glass bottle |
384.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane-tetrahydrofuran complex | |
1M solution in tetrahydrofuran, stabilized | |
C4H11BO | |
0.96 to 1.08M | |
0.8760g/mL | |
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf | |
RMCYTHFAWCWRFA-UHFFFAOYSA-N | |
oxolane borane | |
11062302 | |
AcroSeal™ Glass bottle | |
Stabilized | |
Liquid |
Colorless | |
14044-65-6, 109-99-9 | |
MFCD00012429 | |
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101 | |
15, 1336 | |
Solubility in water: reacts. | |
B.C1CCOC1 | |
85.94 | |
85.94 | |
0.876 | |
−22°C |
Safety and Handling
GHS H Statement
May cause respiratory irritation. Causes serious eye damage. In contact with water releases flammable gases which may ignite spontaneously. Causes skin irritation. Harmful if swallowed. Highly flammable liquid and vapour. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'einecsNumber' : 237-881-8
missing translation for 'tsca' : TSCA
- Borane-tetrahydrofuran complex, 1M solution in THF, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only