Learn More
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals
Description
It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1860°C |
Melting Point | ∽450°C |
Quantity | 10 kg |
Assay Percent Range | 97.5% min., typically 98.5% |
Solubility Information | Slightly soluble in water and methanol. |
IUPAC Name | oxo[(oxoboranyl)oxy]borane |
Formula Weight | 69.62 |
Sensitivity | Hygroscopic |
Density | 2.46 |
Chemical Name or Material | Boron oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W13K026 | Certificate of Analysis | 19/09/2024 | 012290.A3,012290.36,012290.A9 |
W12I022 | Certificate of Analysis | 16/09/2022 | 012290.A3,012290.36,012290.A9 |
Y16G050 | Certificate of Analysis | 14/09/2022 | 012290.A3,012290.36,012290.A9 |
T10G002 | Certificate of Analysis | 12/04/2022 | 012290.A3,012290.36,012290.A9 |
U25C014 | Certificate of Analysis | 28/12/2021 | 012290.A3,012290.36,012290.A9 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.