missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals

40.60 € - 343.71 €

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo[(oxoboranyl)oxy]borane
SMILES O=BOB=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11368938
View Documents
Thermo Scientific Alfa Aesar
041786.04
2 g
40.60 €
2g
Please to purchase this item. Need a web account? Register with us today!
11378938
View Documents
Thermo Scientific Alfa Aesar
041786.09
10 g
93.67 €
10g
Please to purchase this item. Need a web account? Register with us today!
11358938
View Documents
Thermo Scientific Alfa Aesar
041786.18
50 g
343.71 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-86-2
69.62
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
oxo[(oxoboranyl)oxy]borane
B2O3
MFCD00011315
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
CHEBI:30163
O=BOB=O

Specifications

1860°C
1303-86-2
Powder
B2O3
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
oxo[(oxoboranyl)oxy]borane
518682
69.62
Hygroscopic
Boron oxide
∽450°C
White
99.999%
MFCD00011315
Soluble in water. Slightly soluble in methanol.
O=BOB=O
69.62
CHEBI:30163
Odorless
2.46
Videos
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P201-P202-P281-P308+P313-P501c

H360FD

missing translation for 'einecsNumber' : 215-125-8

missing translation for 'rtecsNumber' : ED7900000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only