missing translation for 'onlineSavingsMsg'
Learn More
Boron trichloride, 1M soln. in hexanes, mixed isomers, Thermo Scientific Chemicals Product Code.: 15418215

Boron trichloride, 1M soln. in hexanes, mixed isomers, Thermo Scientific Chemicals

Product Code. 15418215
100 mL
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 15418215

Brand: Thermo Scientific Alfa Aesar H32833.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A mild, selective combination reagent for the cleavage of primary alkyl aryl ethers. Boron trichloride is a selective demethylating agent. For preparing methyl esters of fatty acids and for transesterification of triglycerides. Mainly used as semiconductor silicon doping source or organic synthesis catalyst, also used for making high purity boron or organic boron.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat. Unstable. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-34-5
Molecular Formula BCl3
Molecular Weight (g/mol) 117.16
MDL Number MFCD00011313
InChI Key FAQYAMRNWDIXMY-UHFFFAOYSA-N
Synonym boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gasShow MoreShow Less
PubChem CID 25135
IUPAC Name trichloroborane
SMILES ClB(Cl)Cl

Specifications

Quantity 100 mL
Linear Formula BCl3
UN Number UN3286
Solubility Information Reacts with water.
Formula Weight 117.17
Concentration or Composition (by Analyte or Components) 1M soln. in hexanes
Sensitivity Moisture sensitive; May form a precipitate
Density 0.738
Chemical Name or Material Boron trichloride, mixed isomers

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251475 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH32833.AP,H32833.AE
Lot Number10241849 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberH32833.AP,H32833.AE
Lot Number10246572 Certificate TypeCertificate of Analysis Date17/05/2023 Catalog NumberH32833.AE
Lot Number10245667 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberH32833.AP,H32833.AE
Lot Number10182298 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32833.AP,H32833.AE
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Boron trichloride, 1M solution in hexanes, mixed isomers
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Hexane, Boron trichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron trichloride, 1M soln. in hexanes, mixed isomers, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.