Learn More
Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 198928000
345.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trichloride | |
1M Solution in Hexane | |
BCl3 | |
MFCD00011313 | |
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
AcroSeal™ Glass bottle | |
800 mL | |
Solution |
Colorless to Yellow | |
10294-34-5, 110-54-3 | |
BCl3 | |
0.9 to 1.1M | |
0.7380g/mL | |
Solubility in water: may decompose. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
0.738 | |
−17°C |
Safety and Handling
- Boron trichloride, 1M solution in hexane
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : Hexane, Boron trichloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only