missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M solution in hexane, AcroSeal™, Thermo Scientific Chemicals

76.48 € - 346.73 €

Specifications

Chemical Name or Material Boron trichloride
Color Colorless to Yellow
Name Note 1M Solution in Hexane
CAS 10294-34-5, 110-54-3
Molecular Formula BCl3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10358240
View Documents
Thermo Scientific Acros
198921000
100 mL AcroSeal™ Glass Bottle
76.48 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11994121
View Documents
Thermo Scientific Acros
198928000
800 mL AcroSeal™ Glass bottle
346.73 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Boron trichloride
1M Solution in Hexane
BCl3
MFCD00011313
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
FAQYAMRNWDIXMY-UHFFFAOYSA-N
trichloroborane
25135
0.738
Solution
Colorless to Yellow
10294-34-5, 110-54-3
BCl3
0.9 to 1.1M
0.7380g/mL
Solubility in water: may decompose.
ClB(Cl)Cl
117.16
117.17
−17°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways. Fatal if swallowed. Fatal if inhaled. Suspected of damaging fertility. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container.

Danger

missing translation for 'einecsNumber' : 233-658-4

missing translation for 'rtecsNumber' : ED1925000

missing translation for 'tsca' : TSCA

Product Identifier
  • Boron trichloride, 1M solution in hexane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P402+P404-Store in a dry place. Store in a closed container.

Supplemental information
  • MIXTURE LIST-Contains : Hexane, Boron trichloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.