Learn More
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 176681000
91.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Boron trichloride | |
1M Solution in Methylene Chloride | |
BCl3 | |
MFCD00011313 | |
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44 | |
15, 1350 | |
Solubility in water: may decompose. | |
ClB(Cl)Cl | |
117.16 | |
117.17 | |
1.326 | |
Liquid |
Colorless to Brown | |
10294-34-5, 75-09-2 | |
BCl3 | |
0.9 to 1.1M | |
1.3260g/mL | |
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas | |
FAQYAMRNWDIXMY-UHFFFAOYSA-N | |
trichloroborane | |
25135 | |
AcroSeal™ Glass bottle | |
100 mL |
Safety and Handling
- Boron trichloride, 1M solution in methylene chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H301-Toxic if swallowed.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only