missing translation for 'onlineSavingsMsg'
Learn More

Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

91.66 € - 360.80 €

Specifications

Chemical Name or Material Boron trichloride
Color Colorless to Brown
Name Note 1M Solution in Methylene Chloride
CAS 10294-34-5, 75-09-2
Molecular Formula BCl3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10011621
View Documents
Thermo Scientific Acros
176681000
100 mL AcroSeal™ Glass bottle
91.66 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10332963
View Documents
Thermo Scientific Acros
176688000
800 mL AcroSeal™ Glass Bottle
360.80 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Boron trichloride
1M Solution in Methylene Chloride
BCl3
MFCD00011313
01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44
15, 1350
Solubility in water: may decompose.
ClB(Cl)Cl
117.16
117.17
Liquid
Colorless to Brown
10294-34-5, 75-09-2
BCl3
0.9 to 1.1M
1.3260g/mL
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
FAQYAMRNWDIXMY-UHFFFAOYSA-N
trichloroborane
25135
1.326
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic if swallowed. May cause drowsiness or dizziness. Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

missing translation for 'einecsNumber' : 233-658-4

missing translation for 'rtecsNumber' : ED1925000

missing translation for 'tsca' : TSCA

Recommended Storage : May darken on storage

Product Identifier
  • Boron trichloride, 1M solution in methylene chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H301-Toxic if swallowed.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.