Learn More
Boron trifluoride, 12% (1.5M) in methanol, Thermo Scientific Chemicals
Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3)
Brand: Thermo Scientific Acros 402761000
30.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent, Lewis acid
Specifications
65.0°C | |
-98.0°C | |
373-57-9, 67-56-1 | |
Solution | |
BF3 | |
MFCD00011316 | |
14, 1349 | |
Solubility in water: may decompose | |
FB(F)F | |
67.81 | |
67.81 | |
Glass bottle | |
0.8700g/mL |
Boron trifluoride | |
1.5M solution in methanol | |
Amber to Colorless | |
100 g | |
BF3 | |
4°C | |
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution | |
WTEOIRVLGSZEPR-UHFFFAOYSA-N | |
trifluoroborane | |
11062313 | |
10 to 15% (w/v BF3) | |
0.87 |
Safety and Handling
- Boron trifluoride, 12% in methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only