Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
Brand: Pyrex 3496/02AS
231.80 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- As 3396 range with intermediate stopcock
- Intermediate stopcock facilitates easy drainage of liquid back into reservoir
- 1.5mm bore PTFE key stopcock
Specifications
10 mL | |
0.02 mL |