Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
Brand:  Pyrex 3496/06AS
250.80 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12842104
  • 271.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
0.1 mL