Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
79.00 € - 158.00 €
Specifications
Material Borosilicate Glass
Stopper Material Polyethylene
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15330895
View Documents
Lenz Laborglasinstrumente™
04000049
250 mL
92.00 €
Pack of 2
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10728731
View Documents
Lenz Laborglasinstrumente™
04000037
100 mL
90.00 €
Pack of 2
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10465601
View Documents
Lenz Laborglasinstrumente™
04000070
1000 mL
158.00 €
Pack of 2
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10475601
View Documents
Lenz Laborglasinstrumente™
04000073
2000 mL
124.00 €
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
15330885
View Documents
Lenz Laborglasinstrumente™
04000058
500 mL
104.00 €
Pack of 2
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
10507551
View Documents
Lenz Laborglasinstrumente™
04000028
50 mL
79.00 €
Pack of 2
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
Description
Description
  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Borosilicate Glass
Polyethylene
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug, without Scale