Learn More
Fisherbrand™ Borosilicate Glass Graduated Cylinder
Measure liquids with blue ceramic scale graduated glass cylindars. The Fisherbrand™ borosilicate glass graduated cylindawith hexagonal base and spout
12.50 € - 65.00 €
Specifications
Material Borosilicate Glass
Class B
Scale Blue Ceramic
View More Specs
Products 9
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
11577702
View Documents
Fisherbrand™
FB55215
1000 mL 10 mL N/A N/A N/A
11547702
View Documents
Fisherbrand™
FB55209
100 mL 1 mL N/A N/A N/A
11517702
View Documents
Fisherbrand™
FB55203
10 mL 0.2 mL
12.75 €
Pack of 2
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
11587702
View Documents
Fisherbrand™
FB55217
2000 mL 20 mL N/A N/A N/A
11557702
View Documents
Fisherbrand™
FB55211
250 mL 2 mL N/A N/A N/A
11527702
View Documents
Fisherbrand™
FB55205
25 mL 0.5 mL N/A N/A N/A
11537702
View Documents
Fisherbrand™
FB55207
50 mL 1 mL N/A N/A N/A
11507702
View Documents
Fisherbrand™
FB55201
5 mL 0.1 mL N/A N/A N/A
11567702
View Documents
Fisherbrand™
FB55213
500 mL 5 mL N/A N/A N/A
Description
Description
  • DIN 12680, BS 604, ISO 4788
  • Graduated in blue ceramic
  • Borosilicate Glass
  • Hexagonal base
  • Spouted
  • Individual reorder code on each product
Specifications
Specifications
Borosilicate Glass
B
Blue Ceramic
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Graduated Cylinder