Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, rimless, heavy wall
Manufactured from borosilicate glass with a heavy wall design for strong resistance to heat and chemicals. Fisherbrand™ Borosilicate Glass Heavy Wall Rimless Test Tubes are ideal for chemical and biological work.
Brand: Fisherbrand™ 90106130
41.97 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for this work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from borosilicate glass 3.3
- Thick-walled and therefore mechanically very resistant
- With round bottom
Specifications
ISO 4142 | |
125 mm | |
16 mm |
14 mL | |
Borosilicate Glass | |
1.8 mm |
Product Content Correction
Product Title
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, rimless, heavy wall > Dimensions: 16 dia. x 125mmH
Spot an opportunity for improvement?Share a Content Correction