Learn More
Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks
Constructed of borosilicate glass. Fisherbrand™ Borosilicate Glass Narrow Neck Erlenmeyer Flasks are conical and graduated.
Brand: Fisherbrand™ SPFS66500-50
48.32 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Graduated
- ISO 1773
- Borosilicate glass
- Conical

Specifications
ISO 1773 | |
50 mL | |
90 mm | |
Yes | |
51 mm |
22 mm | |
Borosilicate Glass | |
Conical | |
Narrow |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.