Learn More
Fisherbrand™ Glass Circle Coverslips
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
Brand:  Fisherbrand™ MCR20019
 View more versions of this product
Product Code. 12323138
  • 30.40 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Specifications
Coverslip
Soda Lime Glass
19 mm
Circle
#1 (0.13 to 0.16 mm)
200/Pk.