Learn More
Fisherbrand™ Glass Circle Coverslips
Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
Brand:  Fisherbrand™ MCR00016
 View more versions of this product
Product Code. 12323148
  • 4.14 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
These Glass Circle Coverslips are available in two thicknesses and a variety of diameters. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Specifications
Coverslip
Soda Lime Glass
16 mm
Circle
#1.5 (0.16 to 0.19 mm)
100 Pk.