Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ 04012037
 View more versions of this product
Product Code. 11882642
  • 75.00 € / Pack of 2
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Scale of approx. volume/mL
  • With screwthread retaining device
Specifications
Specifications
100 mL
Borosilicate Glass
2Pack
Polyethylene