Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
75.00 € - 133.12 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11886204
|
Lenz Laborglasinstrumente™
04012073 |
2000 mL |
130.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11812652
|
Lenz Laborglasinstrumente™
04012070 |
1000 mL |
133.12 €
Pack of 2 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
||||||
11882642
|
Lenz Laborglasinstrumente™
04012037 |
100 mL |
75.00 €
Pack of 2 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11802652
|
Lenz Laborglasinstrumente™
04012058 |
500 mL |
104.00 €
Pack of 2 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11892642
|
Lenz Laborglasinstrumente™
04012049 |
250 mL |
85.00 €
Pack of 2 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
11872642
|
Lenz Laborglasinstrumente™
04012028 |
50 mL |
79.00 €
Pack of 2 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
Description
- With NS socket and PE stopper
- Scale of approx. volume/mL
- With screwthread retaining device
Specifications
Borosilicate Glass | |
Polyethylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale