Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
Brand: Fisherbrand™ FB63132
This item is not returnable.
View return policy
Description
- Borosilicate glass test tubes, grade 3.3
- All ISO 4142
Specifications
ISO 4142 | |
Borosilicate Glass | |
12 mm |
75 mm | |
100 Pack |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Test Tube > Dimensions: 12 dia. x 75mmH
Spot an opportunity for improvement?Share a Content Correction