Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
Brand:  Fisherbrand™ FB63161
 View more versions of this product
Product Code. 11992208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Borosilicate glass test tubes, grade 3.3
  • All ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
125 mm
Borosilicate Glass
16 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Test Tube > Dimensions: 16 dia. x 125mmH