missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Borosilicate Glass Weighing Funnels
Introduce powder via the tubular stem. Fisherbrand™ Borosilicate Glass Weighing Funnels are ideal for weighing small quantities of powder.
Brand: Fisherbrand™ FB51972
Description
Ideal for weighing small quantites of powder. Powder can be introduced via the tubular stem.Specifications
10 mL | |
100 mm | |
Weighing Funnel |
Borosilicate Glass | |
Ideal for weighing small quantities of powder |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Borosilicate Glass Weighing Funnels > Length: 100mm
Spot an opportunity for improvement?Share a Content Correction