Learn More
Tocris Bioscience™ Bosentan
High affinity dual ETA and ETB receptor antagonist;orally bioavailable
Brand:  Tocris Bioscience™ 6232/50
321.38 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18706158
  • 407.00 € / 50mg
Explore more special offers
Description
Description
Bosentan is a high affinity dual ETA and ETB receptor antagonist (Ki values are 4.7 and 95 nM for ETA and ETB, respectively). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C -induced contraction of rat trachea in vitro. Orally bioavailable.
Specifications
Specifications
Antagonist
147536-97-8
551.61g/mol
Non-selective Endothelin Receptor Antagonists
Non-selective Endothelin Receptor
C27H29N5O6S
Store at 4°C