missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Bosentan
High affinity dual ETA and ETB receptor antagonist;orally bioavailable
116.00 € - 407.00 €
Specifications
Product Type | Antagonist |
---|---|
Target Proteins | Non-selective Endothelin Receptor |
CAS | 147536-97-8 |
Molecular Formula | C27H29N5O6S |
Formula Weight | 551.61g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758606
|
Tocris Bioscience™
6232/10 |
10 mg |
116.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706158
|
Tocris Bioscience™
6232/50 |
50 mg |
407.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High affinity dual ETA and ETB receptor antagonist (Ki values are 4.7 and 95 nM for ETA and ETB, respectively). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C-induced contraction of rat trachea in vitro. Orally bioavailable.Specifications
Antagonist | |
147536-97-8 | |
551.61g/mol | |
Non-selective Endothelin Receptor Antagonists |
Non-selective Endothelin Receptor | |
C27H29N5O6S | |
Store at 4°C |
For Research Use Only