missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Chemware™ PFA, Carboy

Closure Material: PTFE

Brand:  Saint-Gobain D1069730

Product Code. 12378238

This item is not returnable. View return policy

Description

Description

  • Chemically inert and ultra-pure; meets USP Class VI criteria
  • Lowest metal ion content of all fluoropolymers
  • Ideal for storage during intermediate phases of pharmaceutical and semiconductor processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.6 gal.
PTFE
Closure
Carboy
10 L
40.5 mm
PFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Saint-Gobain Chemware™ PFA, Carboy > Capacity: 10 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.