missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole Parmer Bottle Holder
Cole Parmer Bottle Holder can be mounted to a wall, hood, or benchtop using attached double-faced tape. Will easily handle most standard wash bottles.
Brand: Cole Parmer 63526-70
69.07 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
Cole Parmer Bottle Holder can be mounted to a wall, hood, or benchtop using attached double-faced tape. Will easily handle most standard wash bottles.
Specifications
Bottle Racks and Carriers | |
White | |
PVC | |
Non-sterile | |
76.2 mm |
Not Autoclavable | |
Bottles (4 x 3-1/8 in. dia.) | |
Mount to wall, hood, or benchtop | |
1/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole Parmer Bottle Holder > bottle holder
Spot an opportunity for improvement?Share a Content Correction