Learn More
Socorex™ Bottle Top Dispenser 520 Model
Brand:  Socorex™ 525.100
 View more versions of this product
Product Code. 13548290
  • 723.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Bottletop Dispenser
EN ISO 8655
Ground Glass plunger
Autoclavable
10 to 100 mL
525