Learn More
Trajan™ BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
Brand:  Trajan™ 054099
 View more versions of this product
Product Code. 10661813
  • 426.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Specifications
370°C
0.1 mm
G27, G36, G41
-40°C to +360/370°C
10 m
Non polar
Pi Bonding with the Aromatic Phenyl Groups
5% Phenyl Polysilphenylene Siloxane
Amines Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC/MS
0.1 μm
BPX5
Fused Silica