Learn More
Trajan™ BPX5 GC Capillary Columns
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
376.00 € - 1480.00 €
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | |||
15218653
|
Trajan™
0541261 |
6 m | 0.32 mm | 1 μm |
417.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10567342
|
Trajan™
054112 |
12 m | 0.22 mm | 0.25 μm |
494.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10442341
|
Trajan™
054119 |
25 m | 0.32 mm | 0.25 μm |
800.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10519452
|
Trajan™
054133 |
12 m | 0.53 mm | 0.25 μm |
510.00 €
Each |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||
12684945
|
Trajan™
054102 |
60 m | 0.25 mm | 0.25 μm |
1215.00 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||
16662612
|
Trajan™
054154 |
60 m | 0.32 mm | 1 μm |
1385.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
16642612
|
Trajan™
054148 |
30 m | 0.53 mm | 1 μm |
890.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
16622612
|
Trajan™
054131 |
25 m | 0.53 mm | 1 μm |
890.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10320941
|
Trajan™
054114 |
50 m | 0.22 mm | 0.25 μm |
1385.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10269190
|
Trajan™
054134 |
25 m | 0.53 mm | 0.25 μm |
890.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15124079
|
Trajan™
0541345 |
30 m | 0.53 mm | 0.5 μm |
890.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10430081
|
Trajan™
054125 |
25 m | 0.32 mm | 0.5 μm |
846.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10268810
|
Trajan™
054113 |
25 m | 0.22 mm | 0.25 μm |
805.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10451031
|
Trajan™
054104 |
25 m | 0.15 mm | 0.25 μm |
690.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10791412
|
Trajan™
054142 |
30 m | 0.22 mm | 0.25 μm |
846.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10208670
|
Trajan™
054146 |
60 m | 0.32 mm | 0.25 μm |
1300.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10567532
|
Trajan™
054118 |
12 m | 0.32 mm | 0.25 μm |
523.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10025450
|
Trajan™
054130 |
12 m | 0.53 mm | 1 μm |
510.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10300711
|
Trajan™
054127 |
12 m | 0.32 mm | 1 μm |
533.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15288643
|
Trajan™
054121 |
15 m | 0.25 mm | 1 μm |
506.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10616021
|
Trajan™
054145 |
30 m | 0.32 mm | 0.25 μm |
805.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10177200
|
Trajan™
054144 |
15 m | 0.32 mm | 0.25 μm |
494.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
12905561
|
Trajan™
054100 |
15 m | 0.25 mm | 0.25 μm |
461.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10762562
|
Trajan™
054126 |
50 m | 0.32 mm | 0.5 μm |
1480.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15298643
|
Trajan™
054122 |
30 m | 0.25 mm | 1 μm |
755.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10661813
|
Trajan™
054099 |
10 m | 0.1 mm | 0.1 μm |
426.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
10055400
|
Trajan™
054129 |
50 m | 0.32 mm | 1 μm |
1405.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15248643
|
Trajan™
054110 |
30 m | 0.15 mm | 0.15 μm |
690.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15238643
|
Trajan™
0541011 |
30 m | 0.25 mm | 0.1 μm |
755.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15228663
|
Trajan™
0542170 |
15 m | 0.25 mm | 0.1 μm |
376.00 €
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
Description
- Very low bleed
- High temperature
- Non-polar
- Ideal for GCMS
- Applications include pesticides, herbicides, hydrocarbons, amines and phenols