Learn More
Brady™ 6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled
6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled is high density ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.
Brand:  Brady™ 066202
126.35 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
 View more versions of this product
Product Code. 11503035
  • 133.00 € / 300m
In Stock
to see stock.
Explore more special offers
Add to basket
Includes:
  • Spindle
Description
Description
  • One-color ribbon for printing a single color onto a label
  • R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Compliance
RoHS compatibility: Pass 2002/95/EC
Specifications
Specifications
RoHS 2002/95/EC
Black
Non-sterile
Spindle
984 ft. x 2.36 in.
984 ft.
2.36 in.
1 in.
IP-R6000HF
Resin
IP300 and IP600 printers
1 Ea.
299.92 m x 59.94 mm (984 ft. x 2.36 in.)
299.92 m
6 cm
2.54 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ 6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled > 59.94mm x 299.92m (2.36 in. x 984 ft.)