Learn More
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
GREENER_CHOICE
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
Brand:  Brady™ 110932
 View more versions of this product
Product Code. 11583894
  • 55.50 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
  • High adhesive
  • Translucent
  • Gloss finish
  • Perfect for autoclaving and for use in below freezing temperatures
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815)
Polyester tape for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL
Black, White
Cartridge (Label)
Label Cartridge
M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™21 LAB Label Printer, BMP™21-PLUS Label Printer
1 Roll
0.048 mm
6.40 m
12.70 mm
6.4 cm
M21-500-461-AW
Acrylic, Permanent
Matte
Polyester
General Identification, Laboratory Identification: Vial, Well Plate
1
-196°C to +110°C
21 ft.
0.5 in.
21 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape > 0.500 in. W; 21 ft. H; Polyester; Black on White