Learn More
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape
GREENER_CHOICE
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
55.50 € - 86.60 €
Specifications
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815)
Adhesive Type Acrylic, Permanent
Color Black, White
Finish Matte
Format Cartridge (Label)
View More Specs
Products 3
Product Code Brand Quantity Height (English) Width (English) Price Quantity & Availability  
Product Code Brand Quantity Height (English) Width (English) Price Quantity & Availability  
12837268
GREENER_CHOICE
View Documents
Brady™
110931
1 Roll 21 ft. 0.3 in.
80.05 €
Each
In Stock
to see stock.
Edge
Add to basket
11583894
GREENER_CHOICE
View Documents
Brady™
110932
1 Roll 21 ft. 0.5 in.
55.50 €
Each
In Stock
to see stock.
Edge
Add to basket
11593894
GREENER_CHOICE
View Documents
Brady™
110933
1 Roll 21 ft. 0.75 in.
86.60 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
  • High adhesive
  • Translucent
  • Gloss finish
  • Perfect for autoclaving and for use in below freezing temperatures
Specifications
Specifications
Halogen-free (DIN VDE 0472/parts 815)
Black, White
Cartridge (Label)
Label Cartridge
M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™21 LAB Label Printer, BMP™21-PLUS Label Printer
21 ft.
6.4 cm
Acrylic, Permanent
Matte
Polyester
General Identification, Laboratory Identification: Vial, Well Plate
1 Roll
21 ft.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, clear polyester film tape