Learn More
Brady™ FreezerBondz II Polyester Laboratory Labels B490
FreezerBondz II Matte White Polyester Laboratory Labels are durable polyester markers designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 142938
 View more versions of this product
Product Code. 17878363
  • 102500.00 € / 1 roll
Explore more special offers
Description
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • White polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Brady Series R4300 ribbon is recommended
Specifications
Specifications
RoHS, 2002/618/EC MCV, 2002895/EC
White
Laboratory Label
For use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
1650/Pk.
0.5 x 0.9 in.
22.86 mm
12.70 mm
Acrylic, Permanent
Polyester
Non-sterile
BBP™33 Printer
0.0021 in.
0.9 in.
0.5 in.