missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hubbell Plugout™

Fits with majority of heavier industrial plug applications

Brand:  Brady™ 065968

Product Code. 10192494

  • 79.35 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in large and small version
  • Yellow

Specifications

Specifications

Clamshell Plug Lockout Device
Yellow
7.00 mm
Industrial plug applications
120.00 mm
1 Ea.
160.27 mm
HUBBELL PLUGOUT - LARGE PLUGOUT
Polypropylene
12.47 x 6.31 x 5.06 in.
For plugs with maximum 4.5 in. dia. and 10.15 in. L with 1.45 in. cord
4
Large
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Hubbell Plugout™ > Large

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.