missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Piston Heads for Titrette™ Bottletop Burette
Replace the piston for the Titrette™ easily in the lab.
Brand: BRAND™ 707530
This item is not returnable.
View return policy
Description
- Fits all serial number ranges
- Available for 10, 25 or 50mL Titrette™ Bottletop Burette
Specifications
Piston Head | |
For 25mL Titrette bottletop burette |