missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Piston Heads for Titrette™ Bottletop Burette

Replace the piston for the Titrette™ easily in the lab.

63.30 €

Specifications

Product Type Piston Head
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10779185
View Documents
BRAND™
707530
For 25mL Titrette bottletop burette
63.30 €
Each
In Stock
to see stock.
Add to basket
 
15673987
View Documents
BRAND™
707531
For 10mL Titrette bottletop burette
63.30 €
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
Description

Description

  • Fits all serial number ranges
  • Available for 10, 25 or 50mL Titrette™ Bottletop Burette
Specifications

Specifications

Piston Head
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.