missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified

Indispensable for preparing dilutions and standard solutions

Brand:  BRAND™ 36978

51.02 EUR valid until 2025-12-31
Use promo code "24375" to get your promotional price.


 View more versions of this product

Product Code. 15188264

  • 53.85 € / Pack of 2
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes with glass stopper with dripping tip
  • Contrast blue enamel print on transparent flasks ensures optimum combination of resistance and readability
  • Available in all ISO sizes from 5 to 2000mL
  • Available in transparent and amber colors
  • Made of low expansion coefficient, chemically resistant borosilicate 3.3 glass
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP
50 mL
A
Transparent
Glass
NS 12/21
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.