Learn More
BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified
Indispensable for preparing dilutions and standard solutions
48.90 € - 141.00 €
Specifications
Certifications/Compliance USP
Material Glass
Class A
View More Specs
Products 16
Product Code Brand Capacity (Metric) Closure Size Color Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Color Price Quantity & Availability  
15158434
View Documents
BRAND™
36968
5 mL NS 10/19 Transparent
48.90 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15192726
View Documents
BRAND™
36977
25 mL NS 10/19 Transparent
49.55 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15188264
View Documents
BRAND™
36978
50 mL NS 12/21 Transparent
53.85 €
Pack of 2
In Stock
to see stock.
Add to basket
15178264
View Documents
BRAND™
36980
200 mL NS 14/23 Transparent
63.65 €
Pack of 2
In Stock
to see stock.
Add to basket
15158264
View Documents
BRAND™
36981
250 mL NS 14/23 Transparent
66.15 €
Pack of 2
In Stock
to see stock.
Add to basket
15891148
View Documents
BRAND™
36982
500 mL NS 19/26 Transparent
78.75 €
Pack of 2
In Stock
to see stock.
Add to basket
15359332
View Documents
BRAND™
36983
1000 mL NS 24/29 Transparent
69.55 €
Each
In Stock
to see stock.
Add to basket
15204069
View Documents
BRAND™
36984
2000 mL NS 29/32 Transparent
92.70 €
Each
In Stock
to see stock.
Add to basket
15613967
View Documents
BRAND™
37461
5 mL NS 10/19 Amber
81.20 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15623967
View Documents
BRAND™
37462
10 mL NS 10/19 Amber
81.20 €
Pack of 2
In Stock
to see stock.
Add to basket
15633967
View Documents
BRAND™
37463
20 mL NS 10/19 Amber
80.80 €
Pack of 2
In Stock
to see stock.
Add to basket
15262845
View Documents
BRAND™
37465
50 mL NS 12/21 Amber
84.70 €
Pack of 2
In Stock
to see stock.
Add to basket
15600327
View Documents
BRAND™
37468
200 mL NS 14/23 Amber
104.00 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15282845
View Documents
BRAND™
37469
250 mL NS 14/23 Amber
113.00 €
Pack of 2
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
15643967
View Documents
BRAND™
37470
500 mL NS 19/26 Amber
141.00 €
Pack of 2
In Stock
to see stock.
Add to basket
15663967
View Documents
BRAND™
37471
1000 mL NS 24/29 Amber
111.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Includes with glass stopper with dripping tip
  • Contrast blue enamel print on transparent flasks ensures optimum combination of resistance and readability
  • Available in all ISO sizes from 5 to 2000mL
  • Available in transparent and amber colors
  • Made of low expansion coefficient, chemically resistant borosilicate 3.3 glass
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
Specifications
Specifications
USP
Glass
A