Learn More
BRAND™ Self-adhesive Plate Sealing Film
Securely seals multiwell plates. Self-adhesive Plate Sealing Film can be applied directly to plates and removed without the use of expensive equipment, or can be applied with a hard rubber roller to ensure secure and uniform attachment of the film to the plate.
Brand:  BRAND™ 701370
175.39 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
 View more versions of this product
Product Code. 10135623
  • 201.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily apply to plates and remove without the use of expensive equipment.
  • Application with a hard rubber roller will ensure secure and uniform attachment of the film to the plate.
  • Sealing films are available in different materials especially suited for storage, immunoassay, or cell and tissue culture:
    • Polyethylene
    • Polyester
    • Aluminum
    • Rayon
    • Polypropylene
    • Vinyl
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyethylene, Polypropylene
50 Sheets
Automation
Sealing Film