missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates
Nontreated sterile microplates for diverse applications
Brand: BRAND™ 781661
130.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- PS surface
- Medium binding
- Sterilized via beta-radiation
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottom available in F- shape
- With lid
- Available in clear, white or black
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
Includes Barcode | |
Standard | |
8 x 12 | |
Clear | |
96 | |
Non-treated | |
Bacteriological assays, screening and for storage |
ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS | |
With Lid | |
Medium Binding | |
Polystyrene | |
Sterile | |
Conical Bottom (V-well) | |
50/Cs. |