missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates
Nontreated sterile microplates for diverse applications
143.00 € - 685.00 €
Specifications
Barcode | Includes Barcode |
---|---|
Certifications/Compliance | ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS |
Format | Standard |
Lid | With Lid |
Array | 8 x 12 |
Product Code | Brand | Well Shape | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Color | Price | Quantity & Availability | ||||
10154183
|
BRAND™
781660 |
Round Bottom (U-well) | Clear |
143.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
10091963
|
BRAND™
781663 |
Rounded Square | Clear |
143.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
10469183
|
BRAND™
781662 |
Flat Bottom (F-well) | Clear |
143.00 €
Pack of 50 |
Estimated Shipment: 11-10-2024
Log in to see stock. |
||||
10616953
|
BRAND™
781661 |
Conical Bottom (V-well) | Clear |
143.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
10479183
|
BRAND™
781668 |
Flat Bottom (F-well) | Black |
257.00 €
Pack of 50 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
10513054
|
BRAND™
781665 |
Flat Bottom (F-well) | White |
257.00 €
Pack of 50 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
10467863
|
BRAND™
781670 |
Flat Bottom (F-well) | White, Clear Bottom |
685.00 €
Pack of 50 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
10657333
|
BRAND™
781671 |
Flat Bottom (F-well) | Black, Clear Bottom |
685.00 €
Pack of 50 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
Description
- PS surface
- Medium binding
- Sterilized via beta-radiation
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottom available in F- shape
- With lid
- Available in clear, white or black
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
Includes Barcode | |
Standard | |
8 x 12 | |
Polystyrene | |
Sterile | |
Bacteriological assays, screening and for storage |
ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS | |
With Lid | |
Medium Binding | |
96 | |
Non-treated | |
50/Cs. |