missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Class A PMP Volumetric Flasks with NS Stoppers

These Class A Volumetric Flasks with NS Stoppers, calibrated to contain, are an excellent break-resistant alternative to glass in the laboratory.

Brand:  BrandTech™ 67404

 View more versions of this product

Product Code. 11308553

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Meet Class A tolerances per DIN EN ISO 1042
  • Manufactured from crystal-clear polymethylpentene
  • Supplied certificate includes mean test value and lot (lot and year imprinted on product)
  • Thermal stress up to 121°C (autoclaving) does not cause permanent exceeding of the tolerance limits
  • However, to preserve markings, cleaning at no higher than 60°C is recommended
  • Available in a range of capacities and stopper sizes
Specifications

Specifications

Calibrated “to contain” per DIN EN ISO 1042
Translucent
PMP
Imprinted lot number, batch certificate
121°C max.
Autoclavable
Polypropylene
250 mL
235 mm
19/26
0.15 mL
NS Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.