Learn More
BrandTech™ Class A PMP Volumetric Flasks with NS Stoppers
These Class A Volumetric Flasks with NS Stoppers, calibrated to contain, are an excellent break-resistant alternative to glass in the laboratory.
112.50 €
Specifications
Calibration Calibrated “to contain” per DIN EN ISO 1042
Closure Material Polypropylene
Color Translucent
Material PMP
Includes Imprinted lot number, batch certificate
View More Specs
Products 1
Product Code Brand Capacity (Metric) Standard Taper Joint Tolerance Price Quantity & Availability  
Product Code Brand Capacity (Metric) Standard Taper Joint Tolerance Price Quantity & Availability  
11308553
View Documents
BrandTech™
67404
250 mL 19/26 0.15 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Meet Class A tolerances per DIN EN ISO 1042
  • Manufactured from crystal-clear polymethylpentene
  • Supplied certificate includes mean test value and lot (lot and year imprinted on product)
  • Thermal stress up to 121°C (autoclaving) does not cause permanent exceeding of the tolerance limits
  • However, to preserve markings, cleaning at no higher than 60°C is recommended
  • Available in a range of capacities and stopper sizes
Specifications
Specifications
Calibrated “to contain” per DIN EN ISO 1042
Translucent
Imprinted lot number, batch certificate
Autoclavable
Polypropylene
PMP
121°C max.
NS Stopper