missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PCR 8-Strip Clear Tubes

Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. PCR 8-Strip Tubes are RNase-, DNase-, and DNA-free and are compatible with most 24-, 32-, 96-, and 384-well thermal blocks.

Brand:  BRAND™ 781332

158.45 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10373173

  • 324.00 € / Pack of 120

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. Tube strips are available with or without caps and can be ordered separately.

  • Thin, uniform walls ensure consistent, precise thermal transfer for excellent results.
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
  • Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
  • Available in convenient tube/cap sets.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
0.2 mL
Yes
Autoclavable
Strip
Strips of 8, with attached individual caps
Polypropylene
Yes
Yes
Tube Strip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number24223 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog Number781332
Lot Number24233 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number781332
Lot Number24273 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number781332
Lot Number24213 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog Number781332
Lot Number24053 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog Number781332
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ PCR 8-Strip Clear Tubes > Tube Strip; w/ individual attached Flat Cap

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.