missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PCR 8-Strip Clear Tubes
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples. PCR 8-Strip Tubes are RNase-, DNase-, and DNA-free and are compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
109.00 € - 324.00 €
Specifications
Material | Polypropylene |
---|---|
Dnase Free | Yes |
RNase Free | Yes |
Autoclavable | Autoclavable |
Format | Strip |
Product Code | Brand | Color | Product Type | Closure Included | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Product Type | Closure Included | Volume (Metric) | Price | Quantity & Availability | ||
10249500
|
BRAND™
781320 |
Clear | Tube Strip | No | 0.2 mL |
117.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10448092
|
BRAND™
781321 |
Rose | Tube Strip | No | 0.2 mL |
109.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10375722
|
BRAND™
781322 |
Yellow | Tube Strip | No | 0.2 mL |
109.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10405982
|
BRAND™
781323 |
Green | Tube Strip | No | 0.2 mL |
109.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10561783
|
BRAND™
781324 |
Blue | Tube Strip | No | 0.2 mL |
109.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10747033
|
BRAND™
781333 |
Clear | Tube Strip | Yes | 0.15 mL |
178.00 €
Pack of 120 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10177080
|
BRAND™
781330 |
Clear | Tube Strip | Yes | 0.2 mL |
161.00 €
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10373173
|
BRAND™
781332 |
Clear | Tube Strip | Yes | 0.2 mL |
324.00 €
Pack of 120 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11668419
|
BRAND™
781326 |
Clear | Tubes and Flat Cap | Yes | 0.2 mL |
295.00 €
Pack of 250 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. Tube strips are available with or without caps and can be ordered separately.- Thin, uniform walls ensure consistent, precise thermal transfer for excellent results.
- Autoclavable
- RNase-, DNase-, and DNA-free.
- Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
- Available in convenient tube/cap sets.
Specifications
Polypropylene | |
Yes | |
Strip |
Yes | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ PCR 8-Strip Clear Tubes