missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Thin Wall 0.2 mL PCR Tubes with Attached Caps
Manufactured without lubricants or chemical additives that can contaminate reagents and samples.Thin Wall 0.2mL PCR Tubes with Attached Caps are autoclavable and RNase-, DNase-, and DNA-free. Thin, uniform walls ensure consistent, precise thermal transfer.
Brand: BRAND™ 781304
60.05 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.- Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
- Autoclavable and RNase-, DNase-, and DNA-free.
- Compatible with most thermal cyclers.
- Supplied with caps that form a tight seal and help prevent sample evaporation, yet are easy to open and close.

Specifications
RNase, DNase and DNA free | |
0.2 mL | |
Thermal cyclers for excellent reproducible results in most PCR applications | |
Polypropylene | |
Autoclavable | |
Single-use | |
0.2 mL |
Blue | |
1000 Pack | |
Non-sterile | |
Flat Cap | |
Yes | |
Tube | |
Thin Wall PCR Tube |
Brand PCR Products
File size: 7.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Thin Wall 0.2 mL PCR Tubes with Attached Caps > 0.2mL; Blue
Spot an opportunity for improvement?Share a Content Correction